missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Zymark™ Rapid Plate/Sciclone and Tecan™ TEMO Acquarius Genmate: 50μL

Product Code. p-4906606 Shop All Axygen Scientific Products
Click to view available options
Tip Style:
Automation, Round
Filtered:
Filtered
Non-filtered
Sterility:
Non-sterile
Sterile
For Use With (Equipment):
Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius MCA96 head, Zymark/Caliper RapidPlate, SciClone, SciClone HV96, CAS-4200-1ch. 20-150μL/sec. head
Unit Size:
Pack of 4800
Pack of 960
This item is not returnable. View return policy

Product Code. 12117860

Brand: Axygen™ ZT50R

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Clear, autoclavable tips; color-coded racks.

  • High throughput screening
  • Volume: 50μL
  • Rack Color: Light Green
  • Color: Clear
  • Designed for use with Zymark™ RapidPlate™, Zymark™ SciClone, Tecan™ Te-MO™, Tecan™ Genmate™ and Tecan™ Aquarius™ automated liquid handling systems
  • Certified RNase, DNase and Pyrogen-Free
  • Format: 96 well

Order Info

96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 50 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Rack Color Light Green
Autoclavable Autoclavable
Certifications/Compliance Non-ANSI
For Use With (Application) Maxymum Recovery™ to further improve accuracy and precision while reducing sample loss
Format Rack
Tip Style Automation, Round
For Use With (Equipment) Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius MCA96 head, Zymark/Caliper RapidPlate, SciClone, SciClone HV96, CAS-4200-1ch. 20-150μL/sec. head
Product Line Caliper Zymark
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.