Learn More
Axygen™ Automation Tips for Zymark™/Caliper, 25 μL
Compatible with automated liquid handling systems. Axygen™ Automation Tips for Zymark™/Caliper, 25μL are non-filtered pipette tips in 384-well racks. Constructed of clear polypropyle under strict controls to ensure quality. Options include sterile and non-sterile tips.
Brand: Axygen™ ZT-384-C-R
Code : A0
Additional Details : Weight : 22.35000kg
Description
- Tip format: 96 tip and 384 tip
- Tip volume range: 25 μL to 200 μL
- Tip material: Clear polypropylene
- RNase-/DNase-/human gDNA-free
- Nonpyrogenic
- Available options:
- With or without aerosol-resistant filters
- Nonsterile or sterile
- Standard or Axygen™ Maxymum Recovery™ surface - Compatible heads: HV96, CAS-4200-1 (20 - 150 μL/sec.), LV96, LV384
- Rack format: Non-ANSI (96) and ANSI/SLAS (384)
- Rack material: Polypropylene
Specifications
30 μL | |
Non-sterile | |
Light Green | |
DNase, RNase Free | |
Rack | |
Non-SBS Rack | |
Zymark/Caliper RapidPlate 96/384, SciClone, Zephyr LV96, LV384 Head |
Non-Filtered | |
Clear | |
Autoclavable | |
Non-pyrogenic | |
Automation, Round | |
50 Racks/Cs. | |
Polypropylene |