missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Automation Micropipette Tips

Product Code. 11384325 Shop All Axygen Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Tip Style:
Automation, Genomic/Wide Orifice
Automation, Maxymum Recovery, Round
Automation, Round
Unit Size:
Pack of 4800
Pack of 960
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Tip Style unitSize
11384325 Sterile Automation, Round Pack of 960
12736679 Sterile Automation, Round Pack of 4800
12716679 Sterile Automation, Maxymum Recovery, Round Pack of 4800
11364325 Sterile Automation, Maxymum Recovery, Round Pack of 960
12033939 Non-sterile Automation, Round Pack of 4800
11374325 Non-sterile Automation, Round Pack of 960
11314335 Sterile Automation, Genomic/Wide Orifice Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 11384325 Supplier Axygen™ Supplier No. ZT200RS

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Axygen™ Automation Micropipette Tips are constructed of polypropylene, and feature a variety of available options to suit your needs.

  • For Tecan™/Freedom EVO&trade, Te-Mo™, Aquarius, Zymark™/Caliper, RapidPlate™, and SciClone™ Systems
  • For 96-well, Non-ANSI and ANSI/SLAS (384) format, number 5 polypropylene racks
  • Available with or without aerosol-resistant filters
  • Available sterile/non-sterile
  • Available in standard or Maxymum Recovery™ style

Order Info

96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Rack Color Blue
Autoclavable Autoclavable
Clean Claims RNase-free, DNase-free, Nonpyrogenic
Format Rack
Tip Style Automation, Round
Packaging 96 tips/rack, 10 racks/pack, 5 packs/case
Quantity 50 Racks/Cs.
Product Type Automation Micropipette Tip
For Use With (Equipment) Tecan/Freedom EVO, Te-Mo, Aquarius, Zymark/Caliper, RapidPlate, and SciClone Systems
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.