missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- For Tecan™/Freedom EVO&trade, Te-Mo™, Aquarius, Zymark™/Caliper, RapidPlate™, and SciClone™ Systems
- For 96-well, Non-ANSI and ANSI/SLAS (384) format, number 5 polypropylene racks
- Available with or without aerosol-resistant filters
- Available sterile/non-sterile
- Available in standard or Maxymum Recovery™ style
Order Info
96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Specifications
Specifications
| Volume (Metric) | 200 μL |
| Filtered | Non-filtered |
| Sterility | Sterile |
| Color | Clear |
| Rack Color | Blue |
| Autoclavable | Autoclavable |
| Clean Claims | RNase-free, DNase-free, Nonpyrogenic |
| Format | Rack |
| Tip Style | Automation, Maxymum Recovery, Round |
| Packaging | 96 tips/rack, 10 racks/pack, 5 packs/case |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?