missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Automation Micropipette Tips, PerkinElmer/MultiPROBE Janus™ Systems
GREENER_CHOICE

Product Code. 12792916 Shop All Axygen Scientific Products
Change view
Click to view available options
Volume (Metric):
50 μL
200 μL
Unit Size:
Pack of 4800
Pack of 960
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
12792916 50 μL Pack of 4800
12712916 200 μL Pack of 4800
11310295 200 μL Pack of 960
11330295 50 μL Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12792916 Supplier Axygen™ Supplier No. RRP50CR
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Axygen™ Automation Micropipette Tips, PerkinElmer/MultiPROBE Janus™ Systems are constructed of polypropylene, and feature a variety of available options to suit your needs.

  • Available with or without aerosol-resistant filters
  • Available sterile/non-sterile
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 50 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Clean Claims RNase-free, DNase-free, human gDNA-free, Nonpyrogenic
Format Rack
Tip Style Automation, Round
Packaging 10 racks/pk, 5 pk/cs
Product Type Automation Micropipette Tip
For Use With (Equipment) PerkinElmer MultiProbe
Product Line Tip
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.