missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Automation Micropipette Tips, PerkinElmer/MultiPROBE Janus™ Systems
Axygen™ Automation Micropipette Tips, PerkinElmer/MultiPROBE Janus™ Systems are constructed of polypropylene, and feature a variety of available options to suit your needs.
£135.84 - £820.00
Specifications
Filtered | Non-Filtered |
---|---|
Sterility | Non-sterile |
Color | Clear |
Clean Claims | RNase-free, DNase-free, human gDNA-free, Nonpyrogenic |
Format | Rack |
Product Code | Brand | Volume (Metric) | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | For Use With (Equipment) | Price | Quantity & Availability | ||||
12712916
|
Axygen™
RRP-200-C-R |
200 μL | PerkinElmer MultiProbe |
£820.00
Pack of 4800 |
Estimated Shipment: 29-07-2024
Log in to see stock. |
||||
12792916
|
Axygen™
RRP-50-C-R |
50 μL | PerkinElmer MultiProbe |
£820.00
Pack of 4800 |
Estimated Shipment: 22-07-2024
Log in to see stock. |
||||
11330295
|
Axygen™
RRP-50-C-R |
50 μL | PerkinElmer MultiProbe |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11310295
|
Axygen™
RRP-200-C-R |
200 μL | PerkinElmer MultiProbe |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Available with or without aerosol-resistant filters
- Available sterile/non-sterile
Specifications
Non-Filtered | |
Clear | |
Rack | |
10 racks/pk, 5 pk/cs | |
PerkinElmer MultiProbe | |
Polypropylene |
Non-sterile | |
RNase-free, DNase-free, human gDNA-free, Nonpyrogenic | |
Automation, Round | |
Automation Micropipette Tip | |
Tip |