missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ PerkinElmer™ Janus™ & Multiprobe Robotic Tips
GREENER_CHOICE

Product Code. 10270310 Shop All Axygen Scientific Products
Change view
Click to view available options
Volume (Metric):
20 μL
25 μL
50 μL
175 μL
200 μL
Sterility:
Non-sterile
Sterile
Color:
Black
Clear
Unit Size:
Pack of 4800
Pack of 960
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility Color unitSize
10270310 200 μL Sterile Clear Pack of 4800
12766719 175 μL Sterile Clear Pack of 4800
11350285 175 μL Sterile Clear Pack of 960
12545477 200 μL Sterile Clear Pack of 960
11380285 25 μL Sterile Black Pack of 960
12776719 25 μL Sterile Black Pack of 4800
12772916 50 μL Non-sterile Black Pack of 4800
11340295 50 μL Non-sterile Black Pack of 960
12792906 200 μL Non-sterile Black Pack of 4800
11320295 200 μL Non-sterile Black Pack of 960
12786719 25 μL Sterile Clear Pack of 4800
11370285 25 μL Sterile Clear Pack of 960
12756719 175 μL Sterile Black Pack of 4800
11360285 175 μL Sterile Black Pack of 960
12585477 20 μL Sterile Clear Pack of 960
10308809 20 μL Sterile Clear Pack of 4800
12752916 20 μL Non-sterile Clear Pack of 4800
11390285 20 μL Non-sterile Clear Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
This item is not returnable. View return policy
Product Code. 10270310 Supplier Axygen™ Supplier No. RRP200CRS
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Volume ranges from 20 to 200μL

  • Racked
  • Black and Clear tips available
  • With Liquid level sensing
  • Nonsterile and Presterilized tips
  • Blue, Red and Green rack color inserts

Order Info

96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Clean Claims DNase, RNase, and Pyrogen free
Format Rack
Tip Style Automation, Round
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) PerkinElmer Janus, PerkinElmer MultiProbe Variospan-1, 4, 8 Head
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.