missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Racked
- Black and Clear tips available
- With Liquid level sensing
- Nonsterile and Presterilized tips
- Blue, Red and Green rack color inserts
Order Info
96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Specifications
Specifications
| Volume (Metric) | 20 μL |
| Filtered | Non-filtered |
| Sterility | Non-sterile |
| Color | Clear |
| Rack Color | Blue |
| Autoclavable | Autoclavable |
| Clean Claims | DNase, RNase, and Pyrogen free |
| Format | Rack |
| Tip Style | Automation, Round |
| Packaging | SBS Rack |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?