Learn More
Axygen™ Maximum Recovery™ Polypropylene Pipette Tip
For ultra-low retention and high yield
Brand:  Axygen™ T-200-Y-R
Code : A0
Additional Details : Weight : 1.28880kg
 View more versions of this product
Product Code. 12679595
  • £49.60 / Pack of 960
Estimated Shipment: 26-09-2024
to see stock.
Add to basket
Description
Description
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Beveled
  • Maxymum Recovery technology for ultra-low retention and high yield
  • Sterile
Specifications
Specifications
200 μL
Yellow
Used with single and multichannel pipettes
Universal Fit
Polypropylene
Sterile
DNase, RNase, and Pyrogen Free
Rack
Maximum Recovery