Learn More
Axygen™ GentleFit™ Dual-Shot Filter Pipet Tips
Soft, leakproof seal provides a universal fit
£615.47 - £697.00
Specifications
Filtered | Filtered |
---|---|
Sterility | Sterile |
Format | Rack |
Material | Polypropylene |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
12722786
|
Axygen
TLF-1000-R-S |
1000 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12712786
|
Axygen
TLF-200-R-S |
200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12702786
|
Axygen
TLF-10-R-S |
10 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
These filter tips are manufactured from two different resins. The thermoplastic elastomer (TPE) top is easy to seal, provides a soft eject, and is colored-coded by volume. The rigid, polypropylene body ensures straightness, affords glass-like clarity and is non-reactive and graduated.
- Versatile; dual-shot technology standardizes most research-grade pipetters
- Available in three sizes: 10, 200 and 1000μL
- Reduced sealing force
- Minimal reciprocal ejection force
- Economical; imposes less wear on pipetters
- Ergonomic design; soft tip requires less pressure to create an optimum seal
- Ideal for multichannel pipetters
- Color-coded top identifies volume
- Presterilized
- Certified to be free from RNase, DNase, and human DNA
Specifications
Filtered | |
Sterile | |
Rack | |
Polypropylene |