Learn More
Axygen™ GentleFit™ Dual-Shot Filter Pipet Tips
Soft, leakproof seal provides a universal fit
Brand:  Axygen TLF-1000-R-S
Code : 66
Additional Details : Weight : 10.60000kg
 View more versions of this product
Product Code. 12722786
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
These filter tips are manufactured from two different resins. The thermoplastic elastomer (TPE) top is easy to seal, provides a soft eject, and is colored-coded by volume. The rigid, polypropylene body ensures straightness, affords glass-like clarity and is non-reactive and graduated.
  • Versatile; dual-shot technology standardizes most research-grade pipetters
  • Available in three sizes: 10, 200 and 1000μL
  • Reduced sealing force
  • Minimal reciprocal ejection force
  • Economical; imposes less wear on pipetters
  • Ergonomic design; soft tip requires less pressure to create an optimum seal
  • Ideal for multichannel pipetters
  • Color-coded top identifies volume
  • Presterilized
  • Certified to be free from RNase, DNase, and human DNA
Specifications
Specifications
1000 μL
Sterile
Polypropylene
Filtered
Rack