missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ FLIPR™ Liberty Robotic Tips
GREENER_CHOICE

Product Code. p-4102779 Shop All Axygen Scientific Products
Click to view available options
Color:
Black
Clear
Unit Size:
Pack of 19200
Pack of 3840
This item is not returnable. View return policy

Product Code. 12722876

Brand: Axygen™ LT384R

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Accurate sample delivery

  • Available in clear and black
  • Volume: 30μL
  • Available as racked or racked and pre-sterilized
  • For use with Molecular Devices Liberty head
  • Packaging: 384 Tips/rack; 10 Racks/unit; 5 Units/Cs.

Certifications

Certified RNase-, DNase- and endotoxin-safe

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 30 μL
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Clean Claims DNase, RNase, and Pyrogen free
Format Rack
Tip Style Automation, Round
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Molecular Devices FLIPR 384-well Head
Product Line FLIPR
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.