missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ FDSS™ Robotic Tips

Product Code. 15581383 Shop All Axygen Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Hamamatsu FDSS 7000 System
Unit Size:
Pack of 19200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15581383 Hamamatsu FDSS 7000 System Pack of 19200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15581383 Supplier Axygen™ Supplier No. FDSS1536BKR
  • ONLINE EXCLUSIVE PRICE
    £7880.00
    £7735.33 / Pack of 19200
    Save £144.67
    1% Off
Estimated Shipment: 11-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For Hamamatsu FDSS series systems

  • For 384-Well plates
  • Tip color: Black
  • Capacity: 50μL
  • Packaging: 384 Tips/rack; 10 Racks/unit; 5 Units/Cs.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 50 μL
Filtered Non-filtered
Sterility Non-sterile
Color Black
Autoclavable Autoclavable
Certifications/Compliance Non-ANSI
Format Rack
Tip Style Automation, Round
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Hamamatsu FDSS 7000 System
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.