missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Biomek™ FX/Wellpro™ Robotic Tips
GREENER_CHOICE

Product Code. 11376294 Shop All Axygen Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Tip Style:
Automation, Extended Length
Extended Length, Automation
Low Retention, Extended Length, Automation
Unit Size:
Pack of 19200
Pack of 3840
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Tip Style unitSize
11376294 Non-sterile Low Retention, Extended Length, Automation Pack of 3840
12732866 Non-sterile Low Retention, Extended Length, Automation Pack of 19200
12742866 Sterile Automation, Extended Length Pack of 19200
11386294 Sterile Automation, Extended Length Pack of 3840
11396294 Non-sterile Extended Length, Automation Pack of 3840
12752866 Non-sterile Extended Length, Automation Pack of 19200
12762866 Sterile Extended Length, Automation Pack of 19200
11306304 Sterile Extended Length, Automation Pack of 3840
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 11376294 Supplier Axygen™ Supplier No. FX384XLLR

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For Beckman FX/Wellpro system

  • Tip format: 384 tip
  • Tip volume range: 25 μL to 50 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile 
    - Standard or Axygen™ Maxymum Recovery™ surface
  • Compatible head: AP-384, 384-channel low volume (FXF-384-XL-R-S only)
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 50 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Not Autoclavable
Clean Claims DNase, RNase and Pyrogen free
For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls
Format Rack, 384-well
Tip Style Low Retention, Extended Length, Automation
Packaging 10 Racks/Pk, 5 Pks/Cs.
Quantity 50 Racks/Cs.
For Use With (Equipment) Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Multimek AP384, Dynamic Devices Oasis, Dynamic Devices Oasis LM 384-Channel Low Volume, Hamilton
Product Line Beckman Biomek FX/Wellpro
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.