Learn More
- Home
- Products
- Pipette Products
- Pipette Tips
- Robotic Pipette Tips
- Axygen™ Biomek™ FX/NX Robotic Tips
Axygen™ Biomek™ FX/NX Robotic Tips
Product Code. 11326294 Shop All Axygen Scientific Products| Product Code. | Volume (Metric) | Sterility | Tip Style | Format | unitSize |
|---|---|---|---|---|---|
| 11326294 | 10 μL | Sterile | Automation, Round | Rack, 96 | Pack of 960 |
| 11316294 | 10 μL | Non-sterile | Automation, Round | Rack, 96 | Pack of 960 |
| 12732846 | 10 μL | Non-sterile | Automation, Round | Rack, 96 | Pack of 4800 |
| 12742846 | 10 μL | Sterile | Automation, Round | Rack, 96 | Pack of 4800 |
| 11980948 | 20 μL | Non-sterile | Automation, Round | Rack, 96 | Pack of 960 |
| 12792846 | 20 μL | Non-sterile | Automation, Round | Rack, 96 | Pack of 4800 |
| 12702856 | 20 μL | Sterile | Automation, Round | Rack, 96 | Pack of 4800 |
| 12519177 | 20 μL | Sterile | Automation, Round | Rack, 96 | Pack of 960 |
| 12772846 | 20 μL | Non-sterile | Automation, Round | Rack, 96 | Pack of 4800 |
| 12599167 | 20 μL | Non-sterile | Automation, Round | Rack, 96 | Pack of 960 |
| 12509177 | 20 μL | Sterile | Automation, Round | Rack, 96 | Pack of 960 |
| 12782846 | 20 μL | Sterile | Automation, Round | Rack, 96 | Pack of 4800 |
| 11346294 | 30 μL | Sterile | Automation, Fine Point | Rack, 1536-well | Pack of 3840 |
| 15591383 | 30 μL | Non-sterile | Automation, Fine Point | Rack, 384-well | Pack of 19200 |
| 12722866 | 30 μL | Sterile | Automation, Round | Rack, 384-well | Pack of 19200 |
| 12585727 | 30 μL | Sterile | Automation, Round | Rack, 384-well | Pack of 3840 |
| 12712866 | 30 μL | Non-sterile | Automation, Round | Rack, 384-well | Pack of 19200 |
| 12575727 | 30 μL | Non-sterile | Automation, Round | Rack, 384-well | Pack of 3840 |
| 15501393 | 30 μL | Sterile | Automation, Fine Point | Rack, 384-well | Pack of 19200 |
| 12792856 | 30 μL | Non-sterile | Automation, Round | Rack, 384-well | Pack of 19200 |
| 11356294 | 30 μL | Non-sterile | Automation, Round | Rack, 384-well | Pack of 3840 |
| 12782856 | 45 μL | Sterile | Automation, Fine Point | Rack, 384-well | Pack of 19200 |
| 12545727 | 45 μL | Sterile | Automation, Fine Point | Rack, 384-well | Pack of 3840 |
| 12772856 | 45 μL | Non-sterile | Automation, Fine Point | Rack, 384-well | Pack of 19200 |
| 12535727 | 45 μL | Non-sterile | Automation, Fine Point | Rack, 384-well | Pack of 3840 |
| 12772866 | 50 μL | Non-sterile | Automation, Round | Rack, 96 | Pack of 4800 |
| 12579177 | 50 μL | Non-sterile | Automation, Round | Rack, 96 | Pack of 960 |
| 12782866 | 50 μL | Sterile | Automation, Round | Rack, 96 | Pack of 4800 |
| 12589177 | 50 μL | Sterile | Automation, Round | Rack, 96 | Pack of 960 |
| 12792866 | 50 μL | Non-sterile | Automation, Round | Rack, 96 | Pack of 4800 |
| 12599177 | 50 μL | Non-sterile | Automation, Round | Rack, 96 | Pack of 960 |
| 12509187 | 50 μL | Sterile | Automation, Round | Rack, 96 | Pack of 960 |
| 12702876 | 50 μL | Sterile | Automation, Round | Rack, 96 | Pack of 4800 |
| 12712856 | 250 μL | Non-sterile | Automation, Round | Rack, 96 | Pack of 4800 |
| 12529177 | 250 μL | Non-sterile | Automation, Round | Rack, 96 | Pack of 960 |
| 12722856 | 250 μL | Sterile | Automation, Round | Rack, 96 | Pack of 4800 |
| 12539177 | 250 μL | Sterile | Automation, Round | Rack, 96 | Pack of 960 |
| 11990948 | 250 μL | Non-sterile | Automation, Round | Rack, 96 | Pack of 960 |
| 12732856 | 250 μL | Non-sterile | Automation, Round | Rack, 96 | Pack of 4800 |
| 12742856 | 250 μL | Sterile | Automation, Round | Rack, 96 | Pack of 4800 |
| 12549177 | 250 μL | Sterile | Automation, Round | Rack, 96 | Pack of 960 |
| 12559177 | 250 μL | Non-sterile | Automation, Wide Orifice | Rack, 96 | Pack of 960 |
| 12752856 | 250 μL | Non-sterile | Automation, Wide Orifice | Rack, 96 | Pack of 4800 |
| 12762856 | 250 μL | Sterile | Automation, Wide Orifice | Rack, 96 | Pack of 4800 |
| 12569177 | 250 μL | Sterile | Automation, Wide Orifice | Rack, 96 | Pack of 960 |
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Description
The Axygen brand range of products includes a comprehensive line of tips for Beckman Coulter automated liquid handling systems. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. The pipette tips are also available with MAXYMum Recovery, a unique low-retention technology that improves tip performance and maximizes the use of your precious samples.
- Available in racked, racked sterile and with aerosol barrier (filtered)
- Compatible with Beckman Biomek FX and Beckman Multimek instruments with an AP96 head
- Also compatible with Span-8 pod when Liquid Level Sensing function is turned off
- Unsurpassed straightness to ensure tip enters the well smoothly, every time
- 384/1536-well compatible to meet your most demanding applications
- Wide bore tips are ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
- Fine Point improves accuracy and precision at lower sample volumes
- Maxymum recovery™ tips reduce sample loss, improves reproducibility and accuracy, minimizes usage of precious samples and reagents
Specifications
Specifications
| Volume (Metric) | 10 μL |
| Filtered | Non-filtered |
| Sterility | Sterile |
| Color | Clear |
| Autoclavable | Not Autoclavable |
| Certifications/Compliance | ANSI |
| Clean Claims | DNase, RNase and Pyrogen free |
| For Use With (Application) | Non-pyrogenic, used for stringent specifications under strict process controls |
| Format | Rack, 96 |
| Tip Style | Automation, Round |
| Packaging | 10 Racks/Pk, 5 Pks/Cs. |
| For Use With (Equipment) | Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Biomek 3000, Beckman Coulter Multimek AP96, Beckman Coulter Multimek SPAN-8, Beckman Coulter Multimek MP20, Beckman Coulter Multimek MP200 |
| Product Line | Beckman Biomek |
| Material | Polypropylene |
| Show More Show Less |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.