missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Automation Tips for STARLine, Clear, CO-RE Style Liquid Level Sensing Tip, Sterile
GREENER_CHOICEBIOPHARMA

Product Code. 15753997 Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
10 μL
50 μL
For Use With (Equipment):
Hamilton MICROLAB STAR, Hamilton MICROLAB STARlet, Hamilton MICROLAB STARplus
Hamilton MICROLAB STAR, Hamilton MICROLAB STARlet, Hamilton MICROLAB STARplus, Hamilton MICROLAB Nimbus
Unit Size:
Pack of 11520
This item is not returnable. View return policy

Product Code. 15753997

Brand: Axygen™ HT10CBKSTKS

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For complete compatibility and a consistent fit.

Axygen brand automation tips are engineered to be compatible with original equipment manufacturers' tips, ensuring complete compatibility and a consistent fit. One of the widest product offerings combined with our engineering expertise stringent QA/QC enables you to get optimum performance no matter which automated liquid handler you are working with.
  • Compatible with Hamilton™ Microlab™ STAR, STARlet, STARplus and Nimbus™
  • Conductive black polypropylene
  • Certified DNase-/RNase-free, nonpyrogenic
  • With or without aerosol-resistant filters
  • Nonsterile or sterile
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Sterile
Color Black
Clean Claims DNase, RNase Free
Format Refill
Tip Style Round
For Use With (Equipment) Hamilton MICROLAB STAR, Hamilton MICROLAB STARlet, Hamilton MICROLAB STARplus, Hamilton MICROLAB Nimbus
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.