missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Automation Tips for Agilent™/Velocity11 VPrep™ and Bravo: 384 Tips/Rack
GREENER_CHOICE

Product Code. 15531453 Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
5 μL
10 μL
15 μL
31 μL
50 μL
70 μL
Sterility:
Non-sterile
Sterile
For Use With (Equipment):
Agilent/Velocity VPrep 16ST, Agilent/Velocity VPrep 96ST, Agilent/Velocity VPrep 384ST, Agilent Bravo 384ST
Agilent/Velocity VPrep 384ST, Agilent Bravo 384ST
Unit Size:
Pack of 19200
This item is not returnable. View return policy

Product Code. 15531453

Brand: Axygen™ VT38431LRS

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Comprehensive line of tips for Agilent/Velocity11 automation systems

  • Tip format: 96 tip
  • Tip volume range: 20 μL to 250 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
    - Standard or Axygen™ Maxymum Recovery™ surface
  • Compatible heads: 96LT, 8LT
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene

Order Info

Packaging:10 Racks/Pack, 5 Packs/Case

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 31 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Clean Claims DNase, RNase, and Pyrogen free
For Use With (Application) Maxymum Recovery™ technology for ultra-low retention
Format Rack
Tip Style Automation, Round
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Agilent/Velocity VPrep 16ST, Agilent/Velocity VPrep 96ST, Agilent/Velocity VPrep 384ST, Agilent Bravo 384ST
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.