missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 250 μL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 96 Tip Format
GREENER_CHOICEBIOPHARMA

Product Code. 10221480 Shop All Axygen Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Tip Style:
Low Retention
Standard
Features:
Non-pyrogenic
Non-pyrogenic, Bottom Label
Unit Size:
Pack of 4800
Pack of 960
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Tip Style Features unitSize
10221480 Sterile Standard Non-pyrogenic Pack of 4800
12555567 Non-sterile Standard Non-pyrogenic Pack of 960
10313370 Non-sterile Standard Non-pyrogenic Pack of 4800
12786649 Non-sterile Low Retention Non-pyrogenic Pack of 4800
12535567 Non-sterile Low Retention Non-pyrogenic Pack of 960
12585567 Sterile Standard Non-pyrogenic Pack of 960
12716659 Non-sterile Standard Non-pyrogenic, Bottom Label Pack of 4800
12565567 Non-sterile Standard Non-pyrogenic, Bottom Label Pack of 960
12575567 Sterile Standard Non-pyrogenic, Bottom Label Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 10221480 Supplier Axygen™ Supplier No. VT250RS
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For use with Agilent/Velocity11 automation systems

Tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.

  • Clear polypropylene
  • Maxymum Recovery™ improves accuracy and precision while reducing sample loss
  • Certified RNase-/DNase-free and nonpyrogenic
  • Nonpyrogenic
  • Available in nonsterile or sterile

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 250 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Certifications/Compliance ANSI
Clean Claims DNase, Rnase Free
For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls
Features Non-pyrogenic
Format Rack
Tip Style Standard
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Agilent/Velocity VPrep 8LT, Agilent/Velocity VPrep 96LT, Agilent Bravo 96LT
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.