missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Axl Recombinant Rabbit Monoclonal Antibody (101), PE, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£142.00 - £283.00
Specifications
| Antigen | Axl |
|---|---|
| Clone | 101 |
| Concentration | 100 μg/mL |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30019318
|
Invitrogen™
MA552172 |
25 Tests |
£142.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30019195
![]() |
Invitrogen™
MA552171 |
100 Tests |
£283.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Axl Recombinant Monoclonal antibody specifically detects Axl in Human samples. It is validated for Flow CytometrySpecifications
| Axl | |
| 100 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.5% BSA and 0.03% ProClin 300 | |
| Adhesion-related kinase; AI323647; ARK; AXL; AXL oncogene; AXL receptor tyrosine kinase; AXL transforming sequence/gene; AZF1; JTK11; oncogene AXL; sAXL; soluble AXL; Tyro7; Tyrosine-protein kinase receptor UFO; Ufo; ufo oncogene homolog | |
| Recombinant Human AXL Protein (Met1-Pro449). | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| AXL |
| 101 | |
| Flow Cytometry | |
| PE | |
| Rabbit | |
| Human | |
| P30530 | |
| 558 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title