missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Axl Recombinant Rabbit Monoclonal Antibody (101), PE, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552172
This item is not returnable.
View return policy
Description
AXL is a chronic myelogenous leukemia-associated oncogene that is also associated with colon cancer and melanoma. The AXL protein is a receptor tyrosine kinase that transduces signals from the extracellular matrix into the cytoplasm by binding growth factors like vitamin K-dependent protein growth-arrest-specific gene 6, and is involved in the stimulation of cell proliferation. TAM receptors are also involved in phagocytosis of apoptotic cells. Mice lacking all three TAM receptors have several degenerative phenotypes linked to inefficient removal of apoptotic cells and membranes (e.g., in the retina and the male reproductive tract) and develop a severe autoimmune phenotype akin to systemic lupus erythematosus, including the production of broad spectrum auto-antibodies. In addition, Axl may function as a putative entry receptor for filoviruses. Axl is also used to identify a specific subpopulation of human blood dendritic cells, also referred to as Siglec 6+/Axl+ dendritic cells. Cellular expression of Axl can be upregulated by TLR ligands, such as LPS or poly I:C. Soluble Axl is generated by proteolytic cleavage of the membrane form. Increased plasma levels may be indicative of inflammation and cancer.
Specifications
| Axl | |
| Recombinant Monoclonal | |
| 100 μg/mL | |
| PBS with 0.5% BSA and 0.03% ProClin 300 | |
| P30530 | |
| AXL | |
| Recombinant Human AXL Protein (Met1-Pro449). | |
| 25 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry | |
| 101 | |
| PE | |
| AXL | |
| Adhesion-related kinase; AI323647; ARK; AXL; AXL oncogene; AXL receptor tyrosine kinase; AXL transforming sequence/gene; AZF1; JTK11; oncogene AXL; sAXL; soluble AXL; Tyro7; Tyrosine-protein kinase receptor UFO; Ufo; ufo oncogene homolog | |
| Rabbit | |
| Protein A | |
| RUO | |
| 558 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction