Learn More
Astell Scientific™ AVC002 Pre and Post Simple Vacuum
Product Code. 15791449
Click to view available options
For Use With (Equipment):
Standard Autoclaves with Heaters in The Chamber or Direct Steam Models
Unit Size:
Each
Product Code. 15791449
Brand: Astell Scientific™ AVC002L
Only null left
Add to basket
Add to basket
Ideal for sterilizing porous materials and other loads where air pockets could easily form. Astell Scientific™ AVC002 Pre and Post Simple Vacuum is suitable for standard autoclaves with heaters in the chamber or direct steam models.
  • Suitable for standard autoclaves with the heaters in the chamber, or direct steam models.
  • Cycle begins with a pre vacuum, reducing the chamber pressure and removing the majority of air followed by a free steaming/ air purge stage.
  • After the sterilizing stage is complete, water in the bottom of the chamber is expelled and then a post vacuum takes place, removing any remaining steam from the chamber and cooling the load.
  • Please note: the AVC002 is a simple vacuum and as such does not have the drying capability of the AVC001 and jacket option.
Specifications
For Use With (Application) Sterilizing Porous Materials
For Use With (Equipment) Standard Autoclaves with Heaters in The Chamber or Direct Steam Models
Model Simple
Product Type Autoclave Vacuum