Learn More
Astell Scientific™ AVC001 Advanced Pulsar Vacuum
Description
- Suitable for all units with an integral steam generator (or external steam supply).
- Astell’s most advanced method of air removal.
- Ideal for sterilizing porous materials and other loads where air pockets could easily form.
- Vacuum cycle starts with a pre vacuum, followed by negative and positive pulsing.
- By introducing steam between vacuum pulses, air is forced out of the autoclave chamber under pressure; the best method for removing air from porous loads and discard waste.
- Following the sterilization stage, a post vacuum removes the steam from the chamber.
- In combination with the jacket option, a drying stage then begins to ensure loads are touch dry upon removal.
Specifications
Specifications
| For Use With (Application) | Sterilizing Porous Materials |
| Model | Advanced, pulsar |
| Product Type | Autoclave Vacuum |