Learn More
Axygen™ Automation Tips for PerkinElmer™, 96-Well
GREENER_CHOICE
Product Code. 12772906 Shop All Axygen Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Volume (Metric):
20 μL
50 μL
200 μL
Unit Size:
Pack of 4800
Pack of 960
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Volume (Metric) unitSize
12772906 Non-sterile 50 μL Pack of 4800
11340235 Non-sterile 50 μL Pack of 960
11320225 Non-sterile 20 μL Pack of 960
12712906 Non-sterile 20 μL Pack of 4800
12752896 Non-sterile 200 μL Pack of 4800
11360225 Non-sterile 200 μL Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12772906 Supplier Axygen™ Supplier No. PK50R
Only null left
Add to basket
Add to basket
Ideal for Janus™ MiniTrak, PlateTrak™ and Evolution P3. Axygen™ Automation Tips for PerkinElmer™, 96-Well are constructed of clear polypropylene. Available in sterile/non-sterile, filtered/non-filtered options, from 20 to 200 μL.
Assembled via automation under strict process controls to ensure consistent performance and quality
  • Robotic tips use no mold-releasing agents
  • Nucleic acid, nuclease and endotoxin-safe for complete protection against contamination
  • Tips are certified RNase-/DNase-free and nonpyrogenic
  • Compatible with PerkinElmer™ MDT-96 P235, MDT-96 P50 and MDT-384 30μL head for:
    • Janus™
    • Evolution P3
    • Minitrak
    • PlateTrak™
Multiple options
  • Presterilized and non-sterile tips
  • With or without aerosol-resistant polyethylene filters
  • Wide bore series for working with viscous materials, genomic DNA or delicate cell cultures
  • Available in a range of volumes: 20, 30, 50, 180 and 200μL
Order Info
Rack: 96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Compliance
Axygen products are manufactured in a clean, controlled state of the art facility and certified RNase-, DNase- and endotoxin-safe
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 50 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Rack Color Green
Certifications/Compliance ANSI
Clean Claims DNase, Rnase Free
For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls, nucleic acid, nuclease and endotoxin-safe for complete protection against contamination
Format Rack
Tip Style Automation, Round
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) PerkinElmer Janus, PerkinElmer Evolution P3, PerkinElmer MiniTrak, PerkinElmer PlateTrak, PerkinElmer MDT-96 P235, PerkinElmer MDT-96 P50, PerkinElmer MDT-384 30μL
Material Polypropylene