missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with AP96 Dispense Head
GREENER_CHOICEBIOPHARMA

Product Code. 11801733
Change view
Click to view available options
Volume (Metric):
8 μL
10 μL
20 μL
40 μL
50 μL
130 μL
175 μL
250 μL
Filtered:
Filtered
Non-filtered
Tip Style:
Extended Length
Low Retention
Standard
Wide Bore
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
Unit Size:
Pack of 4800
Pack of 960
25 product options available for selection
Product selection table with 25 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Filtered Tip Style Fit for Purpose unitSize
11801733 130 μL Filtered Standard Sterile and PCR Ready Pack of 4800
15292929 8 μL Filtered Standard Sterile and PCR Ready Pack of 4800
15202949 10 μL Non-filtered Standard Sterile Pack of 4800
15212949 10 μL Non-filtered Standard Certified Quality Pack of 4800
16456939 40 μL Filtered Standard Sterile and PCR Ready Pack of 4800
16466939 50 μL Non-filtered Standard Sterile Pack of 4800
15105393 50 μL Non-filtered Standard Certified Quality Pack of 4800
12578468 50 μL Non-filtered Low Retention Certified Quality Pack of 4800
11861723 20 μL Filtered Standard Sterile and PCR Ready Pack of 4800
11761273 20 μL Filtered Low Retention Sterile and PCR Ready Pack of 4800
11811733 20 μL Non-filtered Standard Sterile Pack of 4800
11741273 20 μL Non-filtered Low Retention Sterile Pack of 4800
11851723 20 μL Non-filtered Standard Certified Quality Pack of 4800
11751273 20 μL Non-filtered Low Retention Certified Quality Pack of 4800
11791273 130 μL Filtered Low Retention Sterile and PCR Ready Pack of 4800
16476939 130 μL Filtered Wide Bore Sterile and PCR Ready Pack of 4800
11881723 250 μL Non-filtered Standard Sterile Pack of 4800
11771273 250 μL Non-filtered Low Retention Sterile Pack of 4800
16486939 250 μL Non-filtered Wide Bore Sterile Pack of 4800
11891723 250 μL Non-filtered Standard Certified Quality Pack of 4800
11781273 250 μL Non-filtered Low Retention Certified Quality Pack of 4800
16496939 250 μL Non-filtered Wide Bore Certified Quality Pack of 4800
11741353 50 μL Filtered Extended Length Sterile Pack of 4800
11515514 175 μL Filtered Standard Sterile Pack of 960
15272289 50 μL Non-filtered Standard Certified Quality Pack of 4800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
25 options
This item is not returnable. View return policy
Product Code. 11801733 Supplier Thermo Scientific™ Supplier No. 919021
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems through the superior tip straightness and low %CV.

Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.

Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384-, and 1536-deep well plates
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 130 μL
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Certifications/Compliance ISO 9001 and ISO 13485
Clean Claims Free of RNase, DNase, DNA, ATP, and endotoxin
Format Rack
Tip Style Standard
Packaging 96 tips/rack, 10 racks/pack, 5 packs/case
Quantity 4800 tips
For Use With (Equipment) Beckman Coulter Liquid Handling Systems
Product Line Beckman
Fit for Purpose Sterile and PCR Ready
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.