missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Automation Tips for Agilent™/Velocity11 and Bravo, 70 μL

Shop All Axygen Scientific Products


Description
- Manufactured to stringent specifications under strict process controls
- Assembled via automation to ensure consistent performance and quality
- Certified RNase-/DNase-free and nonpyrogenic
- 384-well racks of non-filtered pipette 70μL tips
- Compatible with Agilent™/Velocity VPrep™ and Bravo 384ST, 96ST and 16ST head
- Case includes 50 racks (19,200 tips).
Specifications
Specifications
| Volume (Metric) | 70 μL |
| Filtered | Non-filtered |
| Sterility | Sterile |
| Color | Clear |
| Clean Claims | DNase, RNase Free |
| For Use With (Application) | Non-pyrogenic |
| Format | Rack |
| Tip Style | Automation, Round |
| Packaging | SBS Rack |
| Quantity | 50 Racks/Cs. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?