missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Automation Tips for Agilent™/Velocity11 and Bravo, 70 μL
Shop All Axygen Scientific Products

Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 19200
Pack of 3840
Description
- Manufactured to stringent specifications under strict process controls
- Assembled via automation to ensure consistent performance and quality
- Certified RNase-/DNase-free and nonpyrogenic
- 384-well racks of non-filtered pipette 70μL tips
- Compatible with Agilent™/Velocity VPrep™ and Bravo 384ST, 96ST and 16ST head
- Case includes 50 racks (19,200 tips).
Specifications
Specifications
| Volume (Metric) | 70 μL |
| Filtered | Non-filtered |
| Sterility | Sterile |
| Color | Clear |
| Clean Claims | DNase, RNase Free |
| For Use With (Application) | Non-pyrogenic |
| Format | Rack |
| Tip Style | Automation, Round |
| Packaging | SBS Rack |
| Quantity | 50 Racks/Cs. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction