missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Aurora Kinase/Cdk Inhibitor

Product Code. 15447179
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15447179 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15447179 Supplier Thermo Scientific Chemicals Supplier No. J65997.#0
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
This item is not returnable. View return policy

 

Aurora Kinase/Cdk Inhibitor is a broad spectrum inhibitor of cyclin-dependent kinases and aurora kinases including CDK1/Cyclin B, CDK2/Cyclin A, CDK2/Cyclin E, Aurora-A and Aurora-B. It as been shown to induce growth suppression and mitotic defects, these results suggest that Aurora Kinase/Cdk Inhibitor could be useful for cell cycle analysis and therapy of various cancers, including Ewing's sarcoma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Description White
Content And Storage 2°C to 8°C
For Use With (Application) A cell-permeable triazolylsulfonamido compound that acts a reversible, ATP-competitive kinase inhibitor with selectivities towards Aurora kinases
CAS 443797-96-4
Molecular Formula C15H12F2N6O3S.CH3CN
Formula Weight 435.41
Synonym 4-(5-Amino-1-(2,6-difluorobenzoyl)-1H-[1,2,4]triazol-3-ylamino)-benzenesulfonamide;
Sensitivity Light Sensitive
Solubility Soluble in DMSO at 15mg/ml
Product Type Aurora Kinase/Cdk Inhibitor
Form Powder
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.