missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aurora B Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£175.00
Specifications
| Antigen | Aurora B |
|---|---|
| Concentration | 12.5 mg/mL |
| Dilution | Western Blot 1:1000-1:2000, Chromatin Immunoprecipitation, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 60 μg 300 μg lysate, Immunohistochemistry-Paraffin 1:10-1:500, Microarray |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
Aurora B Polyclonal specifically detects Aurora B in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Microarray.Specifications
| Aurora B | |
| Western Blot 1:1000-1:2000, Chromatin Immunoprecipitation, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 60 μg 300 μg lysate, Immunohistochemistry-Paraffin 1:10-1:500, Microarray | |
| Polyclonal | |
| Rabbit | |
| Chromatin Research | |
| 10mM PBS (pH 7.4) with 15mM Sodium Azide | |
| 9212 | |
| Synthetic peptide corresponding to amino acids 1-19 of human Aurora B with C-terminal added cysteine conjugated to KLH. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 12.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| AIK2EC 2.7.11.1, AIM1Aik2, AIM-1STK-1, ARK2ARK-2, AurB, aurkb-sv2, Aurora- and IPL1-like midbody-associated protein 1, aurora kinase BSerine/threonine-protein kinase aurora-B, aurora kinase B-Sv1, aurora kinase B-Sv2, Aurora/IPL1-related kinase 2, aurora-1, aurora-B, Aurora-related kinase 2, EC 2.7.11, IPL1, serine/threonine kinase 12, serine/threonine-protein kinase 12, STK12aurkb-sv1, STK5 | |
| AURKB | |
| IgG | |
| Affinity Purified | |
| Aurora B |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title