missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aurora B Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-294-0.025ml
This item is not returnable.
View return policy
Description
Aurora B Polyclonal specifically detects Aurora B in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Microarray.
Specifications
| Aurora B | |
| Polyclonal | |
| Unconjugated | |
| 10mM PBS (pH 7.4) with 15mM Sodium Azide | |
| AURKB | |
| Synthetic peptide corresponding to amino acids 1-19 of human Aurora B with C-terminal added cysteine conjugated to KLH. | |
| 0.025 mL | |
| Chromatin Research | |
| 9212 | |
| Human | |
| IgG |
| Western Blot | |
| 12.5 mg/mL | |
| Western Blot 1:1000-1:2000, Chromatin Immunoprecipitation, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 60 μg 300 μg lysate, Immunohistochemistry-Paraffin 1:10-1:500, Microarray | |
| AIK2EC 2.7.11.1, AIM1Aik2, AIM-1STK-1, ARK2ARK-2, AurB, aurkb-sv2, Aurora- and IPL1-like midbody-associated protein 1, aurora kinase BSerine/threonine-protein kinase aurora-B, aurora kinase B-Sv1, aurora kinase B-Sv2, Aurora/IPL1-related kinase 2, aurora-1, aurora-B, Aurora-related kinase 2, EC 2.7.11, IPL1, serine/threonine kinase 12, serine/threonine-protein kinase 12, STK12aurkb-sv1, STK5 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Aurora B | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction