missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATXN3L Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | ATXN3L |
|---|---|
| Concentration | 0.65 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16882215
|
Proteintech
23166-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16872215
|
Proteintech
23166-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATXN3L, also named as Ataxin-3-like protein, is a 355 amino acid protein, which contains 1 Josephin domain and 2 UIM (ubiquitin-interacting motif) domains. ATXN3L localizes in the nucleus and as a deubiquitinating enzyme that cleaves both ′Lys-48′-linked and ′Lys-63′-linked poly-ubiquitin chains.Specifications
| ATXN3L | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9H3M9 | |
| 92552 | |
| ATXN3L Fusion Protein Ag19611 | |
| Primary | |
| -20°C | |
| ATXN3L |
| 0.65 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| ataxin 3 like, ATX3L, ATXN3L, MJDL, Putative ataxin 3 like protein | |
| ATXN3L | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title