missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATXN3L Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23166-1-AP-20UL
This item is not returnable.
View return policy
Description
ATXN3L, also named as Ataxin-3-like protein, is a 355 amino acid protein, which contains 1 Josephin domain and 2 UIM (ubiquitin-interacting motif) domains. ATXN3L localizes in the nucleus and as a deubiquitinating enzyme that cleaves both ′Lys-48′-linked and ′Lys-63′-linked poly-ubiquitin chains.Specifications
| ATXN3L | |
| Polyclonal | |
| Unconjugated | |
| ATXN3L | |
| ataxin 3 like, ATX3L, ATXN3L, MJDL, Putative ataxin 3 like protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 92552 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.65 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H3M9 | |
| ATXN3L | |
| ATXN3L Fusion Protein Ag19611 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction