missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATPase Inhibitory Factor 1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £368.00
Specifications
| Antigen | ATPase Inhibitory Factor 1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50-1:100, Immunoprecipitation 1:1000 - 1:5000 |
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18688632
|
Novus Biologicals
NBP2-92891-0.02ml |
0.02 mL |
£161.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18695982
|
Novus Biologicals
NBP2-92891-0.1ml |
0.1 mL |
£368.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATPase Inhibitory Factor 1 Polyclonal antibody specifically detects ATPase Inhibitory Factor 1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
| ATPase Inhibitory Factor 1 | |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS with 50% glycerol, pH7.3. | |
| 93974 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50-1:100, Immunoprecipitation 1:1000 - 1:5000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| ATP synthase inhibitor protein, ATPase inhibitor protein, ATPase inhibitor, mitochondrial, ATPase inhibitory factor 1, ATPIIF(1), ATPIP, IF1, Inhibitor of F(1)F(o)-ATPase, IP, MGC1167, MGC8898 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 26-106 of human ATPIF1 (NP_057395.1). GSDQSENVDRGAGSIREAGGAFGKREQAEEERYFRAQSREQLAALKKHHEEEIVHHKKEIERLQKEIERHKQKIKMLKHDD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title