missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V1G2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | ATP6V1G2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ATP6V1G2 Polyclonal specifically detects ATP6V1G2 in Human samples. It is validated for Western Blot.Specifications
| ATP6V1G2 | |
| Polyclonal | |
| Rabbit | |
| NP_569730 | |
| 534 | |
| Synthetic peptide directed towards the middle region of human ATP6V1G2The immunogen for this antibody is ATP6V1G2. Peptide sequence NLSAEVEQATRRQVQGMQSSQQRNRERVLAQLLGMVCDVRPQVHPNYRIS. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| ATP6G2, ATPase, H+ transporting, lysosomal (vacuolar proton pump), ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G isoform 2, ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G2, Em:AC004181.3, subunit G2, vacuolar ATP synthase subunit G 2, vacuolar proton pump G subunit 2, Vacuolar proton pump subunit G 2, V-ATPase 13 kDa subunit 2, V-ATPase subunit G 2, Vma10, V-type proton ATPase subunit G 2 | |
| ATP6V1G2 | |
| IgG | |
| 13 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title