missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V1C1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92401-0.1ml
This item is not returnable.
View return policy
Description
ATP6V1C1 Polyclonal antibody specifically detects ATP6V1C1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| ATP6V1C1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| ATP6CH+-transporting ATPase chain C, vacuolar, ATP6Dsubunit C of vacuolar proton-ATPase V1 domain, ATPase, H+ transporting, lysosomal (vacuolar proton pump) 42kD, ATPase, H+ transporting, lysosomal 42kDa, V1 subunit C, isoform 1, ATPase, H+ transporting, lysosomal 42kDa, V1 subunit C1, FLJ20057, H(+)-transporting two-sector ATPase, subunit C, vacuolar ATP synthase subunit C, vacuolar proton pump C subunit, Vacuolar proton pump subunit C 1, vacuolar proton pump, 42-kD subunit, vacuolar proton-ATPase, subunit C, VI domain, VATCH+ -ATPase C subunit, V-ATPase C subunit, V-ATPase subunit C 1, Vma5, V-type proton ATPase subunit C 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 300-382 of human ATP6V1C1 (NP_001686.1). LRVFVESVLRYGLPVNFQAMLLQPNKKTLKKLREVLHELYKHLDSSAAAIIDAPMDIPGLNLSQQEYYPYVYYKIDCNLLEFK | |
| 0.1 mL | |
| Endocrinology, Signal Transduction | |
| 528 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction