missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V1B2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | ATP6V1B2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ATP6V1B2 Polyclonal specifically detects ATP6V1B2 in Human, Mouse samples. It is validated for Western Blot.Specifications
| ATP6V1B2 | |
| Polyclonal | |
| Rabbit | |
| P21281 | |
| 526 | |
| IgG | |
| This product is specific to Subunit or Isoform: B, brain isoform. |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 000 subunit, 56/58kD, isoform 2, ATP6B2ATPase, H+ transporting, lysosomal (vacuolar proton pump), beta polypeptide, ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B2, Endomembrane proton pump 58 kDa subunit, H+ transporting two-sector ATPase, HO57ATP6B1B2, vacuolar H+-ATPase 56, Vacuolar proton pump subunit B 2, VATB, V-ATPase B2 subunit, V-ATPase subunit B 2, Vma2, VPP3ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B, isoform 2, V-type proton ATPase subunit B, brain isoform | |
| ATP6V1B2 | |
| Primary | |
| 56 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title