missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V1B2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-54858
This item is not returnable.
View return policy
Description
ATP6V1B2 Polyclonal specifically detects ATP6V1B2 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| ATP6V1B2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| 000 subunit, 56/58kD, isoform 2, ATP6B2ATPase, H+ transporting, lysosomal (vacuolar proton pump), beta polypeptide, ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B2, Endomembrane proton pump 58 kDa subunit, H+ transporting two-sector ATPase, HO57ATP6B1B2, vacuolar H+-ATPase 56, Vacuolar proton pump subunit B 2, VATB, V-ATPase B2 subunit, V-ATPase subunit B 2, Vma2, VPP3ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B, isoform 2, V-type proton ATPase subunit B, brain isoform | |
| Rabbit | |
| 56 kDa | |
| 100 μL | |
| Primary | |
| This product is specific to Subunit or Isoform: B, brain isoform. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P21281 | |
| ATP6V1B2 | |
| Synthetic peptides corresponding to ATP6V1B2(ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B2) The peptide sequence was selected from the middle region of ATP6V1B2. Peptide sequence NFIAQGPYENRTVFETLDIGWQLLRIFPKEMLKRIPQSTLSEFYPRDSAK The peptide sequence for this immunogen was taken from within the described region. | |
| Affinity purified | |
| RUO | |
| 526 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction