Learn More
ATP6V0D1 Mouse, anti-Human, Clone: 1G7A12, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | ATP6V0D1 |
|---|---|
| Clone | 1G7A12 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunoprecipitation, Immunofluorescence, Immunohistochemistry, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18056715
|
Proteintech
68506-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18075505
|
Proteintech
68506-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ATP6V0D1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ATP6V0D1. This antibody recognizes human, mouse, rat, pig, rabbit, chicken, zebrafish antigen. The ATP6V0D1 antibody has been validated for the following applications: IF, IHC, IP, WB, ELISA analysis.
Specifications
| ATP6V0D1 | |
| 1000 μg/mL | |
| Liquid | |
| Human, Mouse, Rat, Pig, Rabbit | |
| 32 kDa accessory protein, ATP6D, ATP6DV, ATP6V0D1, P39, V ATPase AC39 subunit, V ATPase subunit d 1, VATX, VMA6, VPATPD | |
| Primary |
| 1G7A12 | |
| Western Blot, Immunoprecipitation, Immunofluorescence, Immunohistochemistry, ELISA | |
| Mouse | |
| 9114 | |
| IgG1 | |
| Store at -20°C. Stable for one year after shipment. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.