Learn More
ATP6V0D1 Mouse, anti-Human, Clone: 1G7A12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68506-1-IG-20UL
Description
The ATP6V0D1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ATP6V0D1. This antibody recognizes human, mouse, rat, pig, rabbit, chicken, zebrafish antigen. The ATP6V0D1 antibody has been validated for the following applications: IF, IHC, IP, WB, ELISA analysis.
Specifications
| ATP6V0D1 | |
| 1G7A12 | |
| 32 kDa accessory protein, ATP6D, ATP6DV, ATP6V0D1, P39, V ATPase AC39 subunit, V ATPase subunit d 1, VATX, VMA6, VPATPD | |
| 20 μL | |
| 9114 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, Immunoprecipitation, Immunofluorescence, Immunohistochemistry, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig, Rabbit | |
| 150 kDa | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.