missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ATP6AP1 Antibody (3H11B12), Novus Biologicals™
Mouse Monoclonal Antibody
£217.00 - £389.00
Specifications
| Antigen | ATP6AP1 |
|---|---|
| Clone | 3H11B12 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 |
| Applications | Flow Cytometry, ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30032181
|
Novus Biologicals™
NBP3-27175-0.025ml |
0.025 mL |
£217.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30038202
|
Novus Biologicals™
NBP3-27175-0.1ml |
0.1 mL |
£389.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATP6AP1 Monoclonal antibody specifically detects ATP6AP1 in Human samples. It is validated for Flow Cytometry,ELISA,Western BlotSpecifications
| ATP6AP1 | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| 16A, Ac45, ATP6IP1V-ATPase subunit S1, ATP6S1ORF, ATPase, H+ transporting, lysosomal (vacuolar proton pump), subunit 1, ATPase, H+ transporting, lysosomal accessory protein 1, H+ transporting, lysosomal interacting protein 1, H-ATPase subunit, MGC129781, Protein XAP-3, Vacuolar proton pump subunit S1, VATPS1V-ATPase Ac45 subunit, V-type proton ATPase subunit S1, XAP-3, XAP3V-ATPase S1 accessory protein | |
| Purified recombinant fragment of human ATP6AP1 (AA: 51-151) expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 3H11B12 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| 537 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title