missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ATP6AP1 Antibody (3H11B12), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals™ NBP3-27175-0.025ml

 View more versions of this product

Product Code. 30032181

  • £217.00 / 0.03mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ATP6AP1 Monoclonal antibody specifically detects ATP6AP1 in Human samples. It is validated for Flow Cytometry,ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ATP6AP1
Monoclonal
1 mg/mL
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000
16A, Ac45, ATP6IP1V-ATPase subunit S1, ATP6S1ORF, ATPase, H+ transporting, lysosomal (vacuolar proton pump), subunit 1, ATPase, H+ transporting, lysosomal accessory protein 1, H+ transporting, lysosomal interacting protein 1, H-ATPase subunit, MGC129781, Protein XAP-3, Vacuolar proton pump subunit S1, VATPS1V-ATPase Ac45 subunit, V-type proton ATPase subunit S1, XAP-3, XAP3V-ATPase S1 accessory protein
Purified recombinant fragment of human ATP6AP1 (AA: 51-151) expressed in E. Coli.
0.025 mL
Primary
Human
Purified
Flow Cytometry, ELISA, Western Blot
3H11B12
Unconjugated
PBS
Mouse
Protein G purified
RUO
537
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.