missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP4A Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£244.00 - £553.00
Specifications
| Antigen | ATP4A |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18388921
|
Novus Biologicals
NBP3-17134-25UL |
25 μg |
£244.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18399754
|
Novus Biologicals
NBP3-17134-100UL |
100 μg |
£553.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATP4A Polyclonal antibody specifically detects ATP4A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| ATP4A | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| ATP6Agastric H, ATPase, H+/K+ exchanging, alpha polypeptide, ATPase, H+/K+ transporting, alpha polypeptide, EC 3.6.3, EC 3.6.3.10, gastric H, Gastric H(+)/K(+) ATPase subunit alpha, gastric H+/K+ ATPase alpha subunit, gastric hydrogen-potassium ATPase, H(+)-K(+)-ATPase alpha subunit, K-ATPase alpha subunit, K-ATPase catalytic subunit, potassium-transporting ATPase alpha chain 1, Proton pump | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: KKKAGGGGGKRKEKLENMKKEMEINDHQLSVAELEQKYQTSATKGLSASLAAELLLRD | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.2, 40% glycerol | |
| 495 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title