missing translation for 'onlineSavingsMsg'
Learn More

ATP4A Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18399754 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18399754 100 μg 100µL
18388921 25 μg 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18399754 Supplier Novus Biologicals Supplier No. NBP317134100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATP4A Polyclonal antibody specifically detects ATP4A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP4A
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias ATP6Agastric H, ATPase, H+/K+ exchanging, alpha polypeptide, ATPase, H+/K+ transporting, alpha polypeptide, EC 3.6.3, EC 3.6.3.10, gastric H, Gastric H(+)/K(+) ATPase subunit alpha, gastric H+/K+ ATPase alpha subunit, gastric hydrogen-potassium ATPase, H(+)-K(+)-ATPase alpha subunit, K-ATPase alpha subunit, K-ATPase catalytic subunit, potassium-transporting ATPase alpha chain 1, Proton pump
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: KKKAGGGGGKRKEKLENMKKEMEINDHQLSVAELEQKYQTSATKGLSASLAAELLLRD
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 495
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.