Learn More
ATP1B1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15192-1-AP-150UL
 View more versions of this product
Product Code. 16864264
  • £349.00 / 150µL
Description
The protein encoded by this gene belongs to the family of Na+/K+ and H+/K+ ATPases beta chain proteins, and to the subfamily of Na+/K+ -ATPases. Na+/K+ -ATPase is an integral membrane protein responsible for establishing and maintaining the electrochemical gradients of Na and K ions across the plasma membrane. These gradients are essential for osmoregulation, for sodium-coupled transport of a variety of organic and inorganic molecules, and for electrical excitability of nerve and muscle. This enzyme is composed of two subunits, a large catalytic subunit (alpha) and a smaller glycoprotein subunit (beta). The beta subunit regulates, through assembly of alpha/beta heterodimers, the number of sodium pumps transported to the plasma membrane. The glycoprotein subunit of Na+/K+ -ATPase is encoded by multiple genes. This gene encodes a beta 1 subunit. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
Specifications
ATP1B1
Polyclonal
Unconjugated
ATP1B1
ATP1B, ATP1B1, Beta 1 subunit of Na(+) K(+) ATPase, Na+/K+ ATPase beta 1 subunit
Rabbit
Antigen Affinity Chromatography
RUO
11931, 25650, 481
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P05026, P07340, P14094
Atp1b1
ATP1B1 Fusion Protein Ag7279
150 μL
Primary
Human, Mouse, Rat