missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP1A1/2 Mouse, anti-Human, Clone: 3D8G1, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | ATP1A1/2 |
|---|---|
| Clone | 3D8G1 |
| Concentration | 500 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18086895
|
Proteintech
68505-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18087815
|
Proteintech
68505-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ATP1A1/2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ATP1A1/2. This antibody recognizes human, mouse, rat, pig antigen. The ATP1A1/2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ATP1A1/2 | |
| 500 μg/mL | |
| Liquid | |
| Human, Mouse, Rat, Pig | |
| ATP1A1, ATP1A2, FHM2, KIAA0778, MHP2, Sodium pump subunit alpha 2 | |
| Primary |
| 3D8G1 | |
| Western Blot, ELISA | |
| Mouse | |
| 477 | |
| IgG1 | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title