Learn More
ATP1A1/2 Mouse, anti-Human, Clone: 3D8G1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68505-1-IG-20UL
Description
The ATP1A1/2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ATP1A1/2. This antibody recognizes human, mouse, rat, pig antigen. The ATP1A1/2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ATP1A1/2 | |
| 3D8G1 | |
| ATP1A1, ATP1A2, FHM2, KIAA0778, MHP2, Sodium pump subunit alpha 2 | |
| 20 μL | |
| 477 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 500 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| 150 kDa | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.