missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP Citrate Lyase Antibody (5F8D11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£186.00 - £463.00
Specifications
| Antigen | ATP Citrate Lyase |
|---|---|
| Clone | 5F8D11 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:200 - 1:1000, Immunocytochemistry, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18738543
|
Novus Biologicals
NBP2-37429 |
0.1 mL |
£463.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18655575
|
Novus Biologicals
NBP2-37429-0.025ml |
0.025 mL |
£186.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATP Citrate Lyase Monoclonal specifically detects ATP Citrate Lyase in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| ATP Citrate Lyase | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 47 | |
| Purified recombinant fragment of human ACLY (AA: 306-502 ) expressed in E. Coli. | |
| Primary | |
| 125 kDa |
| 5F8D11 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:200 - 1:1000, Immunocytochemistry, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human, Mouse, Rat, Monkey | |
| ACLEC 2.3.3.8, ATP citrate lyase, ATP citrate synthase, ATP-citrate (pro-S-)-lyase, ATP-citrate synthase, ATPCL, Citrate cleavage enzyme, CLATP | |
| ACLY | |
| IgG1 | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title