missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP Citrate Lyase Antibody (5F8D11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37429-0.025ml
This item is not returnable.
View return policy
Description
ATP Citrate Lyase Monoclonal specifically detects ATP Citrate Lyase in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| ATP Citrate Lyase | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:200 - 1:1000, Immunocytochemistry, CyTOF-ready | |
| ACLEC 2.3.3.8, ATP citrate lyase, ATP citrate synthase, ATP-citrate (pro-S-)-lyase, ATP-citrate synthase, ATPCL, Citrate cleavage enzyme, CLATP | |
| Mouse | |
| 125 kDa | |
| 0.025 mL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 5F8D11 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| ACLY | |
| Purified recombinant fragment of human ACLY (AA: 306-502 ) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| 47 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction