missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ATP Assay Kit (Fluorometric)

Product Code. 18639300 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
1 set
Product Code. Quantity unitSize
18639300 96 Tests 1 set
1 options
This item is not returnable. View return policy

Product Code. 18639300

Brand: Novus Biologicals™ NBP324472

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Assay Kit (Fluorometric)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage of components varies. See protocol for specific instructions.
Quantity 96 Tests
Species Non-species specific
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.