missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATOX1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £359.00
Specifications
| Antigen | ATOX1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18612292
|
Novus Biologicals
NBP2-92669-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18602302
|
Novus Biologicals
NBP2-92669-0.1ml |
0.1 mL |
£359.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATOX1 Polyclonal antibody specifically detects ATOX1 in Human samples. It is validated for Western BlotSpecifications
| ATOX1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Golgi Apparatus Markers, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 475 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ATX1, ATX1 (antioxidant protein 1, yeast) homolog 1, ATX1 antioxidant protein 1 homolog (yeast), copper transport protein ATOX1, hah1, HAH1 MGC138453, Metal transport protein ATX1, MGC138455 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-68 of human ATOX1 (NP_004036.1). MPKHEFSVDMTCGGCAEAVSRVLNKLGGVKYDIDLPNKKVCIESEHSMDTLLATLKKTGKTVSYLGLE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title