missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATOX1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92669-0.1ml
This item is not returnable.
View return policy
Description
ATOX1 Polyclonal antibody specifically detects ATOX1 in Human samples. It is validated for Western Blot
Specifications
| ATOX1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| ATX1, ATX1 (antioxidant protein 1, yeast) homolog 1, ATX1 antioxidant protein 1 homolog (yeast), copper transport protein ATOX1, hah1, HAH1 MGC138453, Metal transport protein ATX1, MGC138455 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-68 of human ATOX1 (NP_004036.1). MPKHEFSVDMTCGGCAEAVSRVLNKLGGVKYDIDLPNKKVCIESEHSMDTLLATLKKTGKTVSYLGLE | |
| 0.1 mL | |
| Cancer, Cell Biology, Golgi Apparatus Markers, Signal Transduction | |
| 475 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction